Learn More
3M™ Helmet Integrated Safety Glasses
Click to view available options
Lens Tint:
Clear
Gray
Yellow
Description
- Safety Glasses that easily attach to 3M™ Hard Hats for eye protection
- The integrated eyewear fit closely to the wearer's cheeks, providing excellent protection from particles, dust and approved to EN166
- Durable lens coating helps minimize physical and chemical damage
- Easy to adjust for a good fit
Specifications
Specifications
Certifications/Compliance | EN 166, CE Approved |
Lens Tint | Clear |
Product Type | Safety glasses |
For Use With (Application) | Construction and forestry |
Lens Material | Polycarbonate |