missing translation for 'onlineSavingsMsg'
Learn More

Helios™ Helios Italquartz™ Evaporating Dishes

Brand:  Helios™ 61F00142

 View more versions of this product

Product Code. 11700645

  • 85.10 € / Each
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

55 mL
Evaporating Dish
61 mm
Quartz
27 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers