missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro Heating element spare with mains lead 230V 50/60Hz 1000W Clifton

Brand:  Nickel Electro SA01376

Heating element spare with mains lead 230V 50/60Hz 1000W Clifton

Code : 03

Additional Details : Weight : 0.47000kg

Product Code. 12738868

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers