missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Wide Neck Square Reagent Bottles

Product Code. 11868301
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
  • UN approval for bulk goods

Specifications

Capacity (Metric) 1500 mL
Color Transparent
Diameter (Metric) Inner Neck 69 mm
For Use With (Application) Foodstuffs
Material HDPE
Length (Metric) 108 mm
Width (Metric) 108 mm
Thread Size (Metric) 80 mm
Closure Color Blue
Certifications/Compliance UN Approval
Closure Included Yes
Mouth Wide
Height (Metric) 209 mm
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'