missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huenersdorf™ HDPE Specimen Containers
Secure your items securely in Heunersdorf™ HDPE Specimen Containers, which are made of food-safe polypropylene (PP) for assorted uses.
Specifications
Closure Color | Natural |
---|---|
Closure Material | LDPE |
Closure Type | Hooded Lid |
Color | Natural |
Material | Polyethylene |
Description
- Resistant to acids and chemicals
- Tightly closing lid
- Deep freezable
Specifications
Natural | |
Hooded Lid | |
Polyethylene | |
Deep Freezer, Dishwasher, Microwave | |
Specimen Container |
LDPE | |
Natural | |
Food Safe | |
Stackable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Huenersdorf™ HDPE Specimen Containers