missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC6 Mouse anti-Human, Unconjugated, Clone: 1C7C3, Proteintech
Mouse Monoclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | HDAC6 |
---|---|
Clone | 1C7C3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16869775
|
Proteintech
67250-1-IG-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16859775
|
Proteintech
67250-1-IG-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The HDAC6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC6. This antibody recognizes human antigen. The HDAC6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HDAC6 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
10013 | |
Primary |
1C7C3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human | |
IgG1 |