missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55224-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to HCN4.
HCN4 is a member of the family of hyperpolarization-activated and cyclic-nucleotide-gated (HCN) channels. HCN currents have been linked to pacemaker activity in the heart and brain, resting potential control, as well as neuronal plasticity. It has been shown that HCN4 channels function as receptors for sour taste, and are associated with pacemaker potential generation in the sinoatrial node.Specifications
HCN4 | |
Polyclonal | |
Unconjugated | |
HCN4 | |
HCN4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10021, 330953, 59266 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70507, Q9JKA7, Q9Y3Q4 | |
Hcn4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |