missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hCG alpha Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Specifications
Antigen | hCG alpha |
---|---|
Concentration | 0.36 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16863535
|
Proteintech
25014-1-AP-20UL |
20 μL |
140.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16853535
|
Proteintech
25014-1-AP-150UL |
150 μL |
399.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Luteinizing hormone (LH, also known as lutropin) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-subunit but also a specific beta-subunit, which characterizes each of these hormones.Specifications
hCG alpha | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P01215 | |
1081 | |
hCG alpha Fusion Protein Ag17313 | |
Primary | |
-20°C | |
CGA |
0.36 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CG ALPHA, CGA, Choriogonadotropin alpha chain, Follitropin alpha chain, FSH alpha, FSHA, GPHa, GPHA1, HCG, hCG alpha, LHA, LSH alpha, Lutropin alpha chain, Thyrotropin alpha chain, TSH alpha, TSHA | |
CGA | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |