missing translation for 'onlineSavingsMsg'
Learn More

IKA™ HBR 4 Control Heating Bath

Cylindrical heating bath with an integrated magnetic stirrer to circulate tempering fluid.

Specifications

Controller Type Stepless Heat Control
Capacity (Metric) 4 L
Display LCD
Includes RS 232 interface, external temperature sensor
Height (Metric) 250 mm
View More Specs

Description

Description

  • Cylindrical bath shape
  • Heating elements integrated into the bath base
  • Heat transfer media can be low-viscous oil (50 mPas) or water (H2O)
  • Heat output: 1000 W
  • Two carry handles
  • Infinitely variable safety temperature limiter according to DIN 12877
  • Double-jacketed mantle protects against burns
  • Digital display for target, actual and safety temperatures, and speed
  • Integrated magnetic stirrer drive to circulate the tempering fluid, allowing for better heat distribution in the bath
Specifications

Specifications

Stepless Heat Control
LCD
250 mm
250 mm
4 L
RS 232 interface, external temperature sensor
160 mm
1.4 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.