missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Handle Extension for Unit Dose Sleeve Sampler
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Handle Extension for Unit Dose Sleeve Sampler is ideal for use with a wide range of free flowing powders and granules.
Brand: Sampling Systems™ 1500A-02
This item is not returnable.
View return policy
Description
- For sampling small volume powders and granules
- Chemically resistant 316L stainless steel construction
- Different sample sizes can be taken using the same sampler
- Allows for easy use of sampler
Specifications
Handle Extension | |
Stainless Steel (316L) | |
Unit Dose Sleeve Sampler | |
600 mm |