Learn More
Hamilton™ 700 Series Microliter™ 8100/8200 Autosampler Syringe
Syringe is designed to work with Bruker/Varian 8100/8200 autosamplers
Brand:  Hamilton™ 202880
Product Code. 10676791
  • 1015.00 € / Each
Explore more special offers
Description
Description
Microliter syringe with a stainless steel plunger that is hand-fitted to the glass barrel and is designed for liquid sampling. Syringe has a removable needle (RN) termination. Available in 10μL volume.
Syringe Terminations
  • Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
  • Needle Point Styles
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauge: 26s
    Specifications
    Specifications
    Borosilicate Glass (Barrel), Stainless Steel (Plunger)
    0.005 in.
    0.019 in.
    2 in.
    51 mm
    2000 psi
    50°C
    701
    20°C
    10 μL
    Cone
    Removable
    26s G
    0.13 mm
    0.483 mm
    1.69 in.
    Microliter Syringe
    122°F
    68°F
    5
    Ea.
    43 mm
    Bruker/Varian 8100/8200 Autosamplers