missing translation for 'onlineSavingsMsg'
Learn More

Hafnium telluride, 99.5% (metals basis), Thermo Scientific Chemicals

121.61 € - 616.07 €

Chemical Identifiers

CAS 39082-23-0
Molecular Formula HfTe2
Molecular Weight (g/mol) 433.69
MDL Number MFCD00049947
InChI Key BCMXGUYIVFZMCT-UHFFFAOYSA-N
Synonym hafnium telluride, ditellanylidenehafnium
PubChem CID 90471877
IUPAC Name bis(tellanylidene)hafnium
SMILES [Te]=[Hf]=[Te]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373618
View Documents
Thermo Scientific Alfa Aesar
039223.03
1 g
121.61 €
1g
Please to purchase this item. Need a web account? Register with us today!
11383618
View Documents
Thermo Scientific Alfa Aesar
039223.06
5 g
616.07 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39082-23-0
433.69
BCMXGUYIVFZMCT-UHFFFAOYSA-N
90471877
[Te]=[Hf]=[Te]
HfTe2
MFCD00049947
hafnium telluride, ditellanylidenehafnium
bis(tellanylidene)hafnium

Specifications

39082-23-0
HfTe2
hafnium telluride, ditellanylidenehafnium
[Te]=[Hf]=[Te]
433.69
433.69
(metals basis)
Powder
MFCD00049947
BCMXGUYIVFZMCT-UHFFFAOYSA-N
bis(tellanylidene)hafnium
90471877
99.5%
Hafnium telluride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'einecsNumber' : 254-280-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.