missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium(IV) oxide, 99.99%, (trace metal basis), Thermo Scientific Chemicals
54.15 € - 210.00 €
Chemical Identifiers
CAS | 12055-23-1 |
---|---|
Molecular Formula | HfO2 |
Molecular Weight (g/mol) | 210.49 |
MDL Number | MFCD00003565 |
InChI Key | WIHZLLGSGQNAGK-UHFFFAOYSA-N |
IUPAC Name | hafnium(4+) dioxidandiide |
SMILES | [O--].[O--].[Hf+4] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
12055-23-1 | |
210.49 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
[O--].[O--].[Hf+4] |
HfO2 | |
MFCD00003565 | |
hafnium(4+) dioxidandiide |
Specifications
12055-23-1 | |
Powder | |
HfO2 | |
MFCD00003565 | |
[O--].[O--].[Hf+4] | |
210.49 | |
85% (Hf typical) | |
9.6800g/mL |
White | |
99.99% | |
HfO2 | |
WIHZLLGSGQNAGK-UHFFFAOYSA-N | |
hafnium(4+) dioxidandiide | |
210.49 | |
Trace Metal Basis | |
Hafnium(IV) oxide |
Safety and Handling
Product Identifier
- Hafnium(IV) oxide
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title