Learn More
Guanidine hydrochloride, 98%, Thermo Scientific Chemicals
Guanidine hydrochloride, CAS # 50-01-1, is an ionic buffer component used for nucleic acids (DNA and RNA) and protein isolation.
30.10 € - 276.00 €
Chemical Identifiers
CAS | 50-01-1 |
---|---|
Molecular Formula | CH6ClN3 |
Molecular Weight (g/mol) | 95.53 |
MDL Number | MFCD00013026 |
InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
PubChem CID | 5742 |
ChEBI | CHEBI:32735 |
IUPAC Name | guanidine;hydrochloride |
SMILES | C(=N)(N)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420024
|
Thermo Scientific Alfa Aesar
A13543.30 |
250 g |
30.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410024
|
Thermo Scientific Alfa Aesar
A13543.0B |
1000 g |
74.80 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430024
|
Thermo Scientific Alfa Aesar
A13543.0I |
5000 g |
276.00 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized in the synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Guanidine hydrochloride is the hydrochloride salt of guanidine, an acetylcholine releasing agent
- It is a normal product of protein metabolism and is sometimes used for pyrimidine synthesis
Application
- Guanidine hydrochloride is a powerful denaturant, useful for taking nucleic acids (DNA and RNA) and proteins out of their native conformation
- Guanidine hydrochloride acts as a strong organic base at physiological pH
- It is suitable for use in protein and nucleic acid purification protocols
Chemical Identifiers
50-01-1 | |
95.53 | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
5742 | |
guanidine;hydrochloride |
CH6ClN3 | |
MFCD00013026 | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
CHEBI:32735 | |
C(=N)(N)N.Cl |
Specifications
50-01-1 | |
1.344 | |
MFCD00013026 | |
Hygroscopic | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
guanidine;hydrochloride | |
5742 | |
95.53 | |
Guanidine hydrochloride |
178°C to 185°C | |
CH6ClN3 | |
3591990 | |
144562 | |
Soluble in water and ethanol. | |
C(=N)(N)N.Cl | |
95.53 | |
CHEBI:32735 | |
98% |
Safety and Handling
Causes serious eye irritation, May form combustible dust concentrations in air, Causes skin irritation, Harmful if swallowed or if inhaled
Serious eye damage/eye irritation (category 2), Skin corrosion/irritation (category 2), Acute toxicity (category 4)
missing translation for 'einecsNumber' : 200-002-3
missing translation for 'rtecsNumber' : MF4300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only