missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20133-1-AP-20UL
This item is not returnable.
View return policy
Description
Small G proteins, such as GTPBP5, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 ).Specifications
GTPBP5 | |
Polyclonal | |
Unconjugated | |
MTG2 | |
GTP binding protein 5, GTPBP5, ObgH1, Protein obg homolog 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26164, 296462, 52856 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H4K7 | |
Mtg2 | |
GTPBP5 Fusion Protein Ag14015 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |