missing translation for 'onlineSavingsMsg'
Learn More

Graphite, Pure, Powder, Fisher Chemical

Product Code. 10336090
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7782-42-5
Packaging Plastic powder jar
Molecular Formula C
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
Quantity 1 kg
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.01
PubChem CID 5462310
ChEBI CHEBI:27594
Formula Weight 12.01g/mol
Physical Form Solid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.