Learn More
Grant Instruments™ Rear Connecting Fittings for RC Series High Precision Recirculating Chillers
Fits 9, 12, or 17mm I.D. hoses. Rear Connecting Fittings for RC Series High Precision Recirculating Chillers are supplied in pairs.
Brand:  Grant Instruments™ RC HF12
 View more versions of this product
Product Code. 12787759
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Fits 9, 12, or 17mm I.D. hoses
  • For use with RC Series High Precision Recirculating Chillers
  • Supplied in pairs.
Specifications
Specifications
12mm I.D. Hose for Recirculating chiller
Rear Connecting Fitting