missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Grant Bio™ Adapter Sets for R-12-10 Interchangeable Centrifuge Rotor

Accommodates a variety of vacutainers for the R-12-10 Interchangabl;e Centrifuge Rotor

Brand:  Grant Instruments™ BN-13-75

 View more versions of this product

Product Code. 13459728

  • 222.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Interchangeable centrifuge rotor for 12 x 10 to 15mL tubes
  • Rounded ends, no caps
  • Dimensions: 16mm dia. x 105mm
Specifications

Specifications

2 to 5 mL
75 mm
12/Pk.
13 mm
R-12-10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.