missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Demeter Pipettes

Excellent for precise and accurate transfer of samples in the laboratory. Hirschmann Instruments™ Demeter Pipettes can be used for a wide range of dilution applications.

Brand:  Hirschmann™ 1500144

Product Code. 13147929

  • 321.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With blue graduation
  • Acc. to DIN 12 838 and 12 839
Specifications

Specifications

For dilution after demeter
Blue
1, 2, 2.1, 2.2 mL
Graduated Pipet
Yes
25 Pack
250 ±5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ Demeter Pipettes > Length: 250 plus-minus 5mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.