missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 608 Joseph Paper

Fisherbrand™ Grade 608 Joseph Paper is very absorbent and no fluffy.

Brand:  Fisherbrand™ 0608M00001

 View more versions of this product

Product Code. 11763913

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Thin paper 25g/m2
  • very absorbent and no fluffy
Applications:
  • Cleaning and drying of, glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals
  • Specifications

    Specifications

    500 mm
    500 x 350 mm
    Cleaning and drying of, Glass containers, tubes, flasks, microscopic plates, bottles in laboratories and hospitals
    608
    350 mm
    Folded sheet
    800 Pk.
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.