missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Grade 1107 Cellulose Technical Filter Paper: Discs
Used for filtration of semi fine particles
Specifications
Material | Cellulose |
---|---|
Particle Retention | 20 to 30μm |
Pore Size | 20 to 30 μm |
For Use With (Application) | For Serum, Culture Medium, dyes, tinctures, vegetal extracts, turbid liquids, juices, Eaux de Cologne, perfumes, essences, hair tonics |
Grade | 1107 |
Description
- Thick smooth paper with medium pore structure
Specifications
Cellulose | |
20 to 30 μm | |
1107 | |
0.38 mm |
20 to 30μm | |
For Serum, Culture Medium, dyes, tinctures, vegetal extracts, turbid liquids, juices, Eaux de Cologne, perfumes, essences, hair tonics | |
100 Pack |