Learn More
Fisherbrand™ Grade 1103 Cellulose Technical Filter Paper
Suitable for removal of fine particles
Brand:  Fisherbrand™ 1103A00017
 View more versions of this product
Product Code. 11749995
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Thick smooth paper
  • Wet strengthened
Specifications
Specifications
Disc
12 to 16μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
110 mm
Cellulose
12 to 16 μm
1103
0.3 mm
110 mm