Learn More
Fisherbrand™ Grade 1103 Cellulose Technical Filter Paper
Suitable for removal of fine particles
Brand:  Fisherbrand™ 1103P00010
 View more versions of this product
Product Code. 11720026
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Thick smooth paper
  • Wet strengthened
Specifications
Specifications
Folded Disc
12 to 16μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
800 mm
Cellulose
12 to 16 μm
1103
0.3 mm
800 mm