missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GR repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23978-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is used to detect the GR repeated sequence. The recombit proteins GST-SNRPD1 and 6xHis tag- SNRPD1 which contains the sequence 'GRGRGRGRGRGRGRGRGR' were used to detect this antibody. Both proteins can be recognized by the antibod
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
GR repeat | |
Polyclonal | |
Unconjugated | |
C9RAN, GGGCC, GR repeat, GRGRGRGRG, GRGRGRGRGR, Poly-GR | |
Peptide | |
20 μL | |
Primary | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction