missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16897-1-AP-20UL
This item is not returnable.
View return policy
Description
Glutamate-pyruvate transaminase. See ALTQTL1 and ALTQTL2 for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.Specifications
GPT | |
Polyclonal | |
Unconjugated | |
GPT | |
AAT1, Alanine aminotransferase 1, ALT1, GPT, GPT 1, GPT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2875, 76282, 81670 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25409, P42867, Q9H3H5 | |
GPT | |
GPT Fusion Protein Ag10424 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction