missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24494-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is used to detect the GP repeated sequence. The recombit protein 6xHis tag-SMARCA4 or GST tag-SMARCA4 which contains the sequence 'GPGPGPGPG' were used to detect this antibody.
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
GP repeat | |
Polyclonal | |
Unconjugated | |
C9RAN, GGGCC, GP repeat, GPGPGPGP, poly GP, poly PG | |
Peptide | |
20 μL | |
Primary | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction