Learn More
Gosselin™ Square HDPE Bottles
Pre-dosed with sodium thiosulfate which inhibits the action of chlorine, bromine and ozone dissolved in water. Gosselin™ Square HDPE Bottles are manufactured from translucent HDPE.
141.06 € - 197.43 €
Specifications
Color Translucent
Material HDPE
Closure Included Yes
Shape Square
View More Specs
Products 6
Product Code Brand Closure Color Diameter (Metric) Inner Neck Sterility Price Quantity & Availability  
Product Code Brand Closure Color Diameter (Metric) Inner Neck Sterility Price Quantity & Availability  
11703094
View Documents
Gosselin™
FC510-02
Red 29 mm Sterile
175.32 €
Pack of 100
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11783084
View Documents
Gosselin™
FC520-02
Red 22 mm Sterile
161.21 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
11793084
View Documents
Gosselin™
FC520-06
Red 22 mm Non-sterile
169.27 €
Pack of 100
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
11740485
View Documents
Gosselin™
FC1000-52
Red 28 mm Sterile
197.43 €
Pack of 77
In Stock
to see stock.
Edge
Add to basket
11865953
View Documents
Gosselin™
FC510-03
White 29 mm Sterile
157.18 €
Pack of 100
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
11723293
View Documents
Gosselin™
FC520-10
White 22 mm Non-sterile
141.06 €
Pack of 100
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
Description
Description
  • Break resistant
  • With molded graduations
  • With either a screw cap with shaped seal or hinged cap with inner lip and safety ring
  • Sterilized by irradiation with peel-away sterilization certificate on the outer carton
Specifications
Specifications
Translucent
HDPE
Yes
Square