Learn More
Gold powder, spherical, APS 0.5-0.8 micron, 99.96+% (metals basis), Thermo Scientific Chemicals
Form: S.A. 0.45-0.70 m2/g
72.06 € - 1370.82 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11425460
|
Thermo Scientific Alfa Aesar
044636.MD |
250 mg |
72.06 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405460
|
Thermo Scientific Alfa Aesar
044636.03 |
1 g |
288.44 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415460
|
Thermo Scientific Alfa Aesar
044636.06 |
5 g |
1370.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3Show More | |
CHEBI:30050 | |
[Au] |
Specifications
Golden | |
Gold colored powder | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Spherical Powder |
7440-57-5 | |
>99.96% (Metals basis) | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
Odorless | |
Gold |
Safety and Handling
missing translation for 'einecsNumber' : 231-165-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.