missing translation for 'onlineSavingsMsg'
Learn More

Gold Nickel wire, 0.5mm (0.02in) dia, annealed, 99.85% (metals basis), Thermo Scientific Chemicals

150.75 € - 819.08 €

Specifications

Chemical Name or Material Gold Nickel wire
Name Note 0.5mm (0.02 in.) dia., Annealed, Temper: soft
Avg. Mol. Wt. or Mol. Wt. Range Au:Ni; 82:18 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD02091707
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397908
View Documents
Thermo Scientific Alfa Aesar
036451.BS
10 cm
150.75 €
10cm
Please to purchase this item. Need a web account? Register with us today!
11307918
View Documents
Thermo Scientific Alfa Aesar
036451.BW
50 cm
608.03 €
50cm
Please to purchase this item. Need a web account? Register with us today!
11317918
View Documents
Thermo Scientific Alfa Aesar
036451.G1
1 m
819.08 €
1m
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gold Nickel wire
Au:Ni; 82:18 wt%
MFCD02091707
≈0.32g/10cm
0.5mm (0.02 in.) dia., Annealed, Temper: soft
(metals basis)
99.85%
Wire
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350-H372

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only