Learn More
GMPPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15517-1-AP-20UL
 View more versions of this product
Product Code. 16834724
  • 145.00 € / 20µL
Explore more special offers
Description
Description
GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.
Specifications
Specifications
GMPPA
Polyclonal
Unconjugated
GMPPA
GMPPA
Rabbit
Antigen Affinity Chromatography
RUO
29926, 501167, 69080
-20°C
Liquid
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5XIC1, Q922H4, Q96IJ6
Gmppa
GMPPA Fusion Protein Ag7789
20 μL
Primary
Rat, Human, Mouse