Learn More
Glycolic acid 70% in water, Thermo Scientific Chemicals
22.01 € - 350.75 €
Chemical Identifiers
CAS | 79-14-1, 7732-18-5 |
---|---|
Molecular Formula | C2H4O3 |
Molecular Weight (g/mol) | 76.051 |
MDL Number | MFCD00004312 |
InChI Key | AEMRFAOFKBGASW-UHFFFAOYSA-N |
Synonym | glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czechShow More |
PubChem CID | 757 |
ChEBI | CHEBI:17497 |
IUPAC Name | 2-hydroxyacetic acid |
SMILES | C(C(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10425851
|
Thermo Scientific Acros
411032500 |
250 g | Glass bottle |
22.01 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10475271
|
Thermo Scientific Acros
411030010 |
1 kg | Glass bottle |
33.87 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935081
|
Thermo Scientific Acros
411030250 |
25 kg | Metal drum |
350.75 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
79-14-1, 7732-18-5 | |
76.051 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
757 | |
2-hydroxyacetic acid |
C2H4O3 | |
MFCD00004312 | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czechShow More | |
CHEBI:17497 | |
C(C(=O)O)O |
Specifications
79-14-1, 7732-18-5 | |
113.0°C | |
C2H4O3 | |
MFCD00004312 | |
AEMRFAOFKBGASW-UHFFFAOYSA-N | |
2-hydroxyacetic acid | |
757 | |
76.04 | |
1.2700 g/mL | |
1.27 |
10.0°C | |
Yellow | |
CH2OHCOOH | |
glycolic acid, hydroxyacetic acid, glycollic acid, hydroxyethanoic acid, acetic acid, hydroxy, glycolate, caswell no. 470, alpha-hydroxyacetic acid, kyselina glykolova, kyselina glykolova czech | |
C(C(=O)O)O | |
76.051 | |
CHEBI:17497 | |
70% | |
Solution | |
Glycolic acid, 70% in water |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May damage fertility. May damage the unborn child.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Danger
missing translation for 'einecsNumber' : 201-180-5
missing translation for 'rtecsNumber' : MC5250000
missing translation for 'tsca' : TSCA
- Glycolic acid, 67% in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.