missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
31.36 € - 443.00 €
Chemical Identifiers
CAS | 5680-79-5 |
---|---|
Molecular Formula | C3H8ClNO2 |
Molecular Weight (g/mol) | 125.55 |
MDL Number | MFCD00012870 |
InChI Key | COQRGFWWJBEXRC-UHFFFAOYSA-N |
Synonym | glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride |
PubChem CID | 122755 |
IUPAC Name | methyl 2-aminoacetate;hydrochloride |
SMILES | [H+].[Cl-].COC(=O)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10255250
|
Thermo Scientific Acros
120101000 |
100 g | Plastic bottle |
31.36 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10510931
|
Thermo Scientific Acros
120105000 |
500 g | Plastic bottle |
112.56 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498318
|
Thermo Scientific Acros
120100025 |
2.5 kg | Plastic bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5680-79-5 | |
125.55 | |
COQRGFWWJBEXRC-UHFFFAOYSA-N | |
122755 | |
[H+].[Cl-].COC(=O)CN |
C3H8ClNO2 | |
MFCD00012870 | |
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride | |
methyl 2-aminoacetate;hydrochloride |
Specifications
5680-79-5 | |
White | |
98% | |
H2NCH2CO2CH3·HCl | |
04,34 | |
Solubility in water: >1000g/L water (20 c) | |
[H+].[Cl-].COC(=O)CN | |
125.55 | |
125.56 | |
Crystals or Powder |
173°C to 177°C | |
Authentic | |
C3H8ClNO2 | |
MFCD00012870 | |
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride | |
COQRGFWWJBEXRC-UHFFFAOYSA-N | |
methyl 2-aminoacetate;hydrochloride | |
122755 | |
98% | |
Glycine methyl ester hydrochloride, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 227-139-1
Product Identifier
- Glycine methyl ester hydrochloride
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title