missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, >99%, Highly Purified, MP Biomedicals™

Product Code. 11421112
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Density 1.2613 g/cu cm at 20°C (Lit.)
Assay Percent Range >99%
Odor Odorless
Quantity 1 L
Formula Weight 92.09
Grade Highly Purified
Physical Form Syrupy Liquid
Chemical Name or Material Glycerol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'