missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15637890
Click to view available options
Quantity:
500 mL
1 L
2 L
4 L
4 x 4 L
Packaging:
Glass bottle
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.25g/mL
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Ignition Point 698°F
Odor Odorless
Packaging Poly bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Solubility Information Completely miscible in water
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'