missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ≥99.0% (GC), Reagent Grade, Honeywell™

Product Code. 15617890
Click to view available options
Quantity:
500 mL
1 L
1 gal.
5 L
20 L
Packaging:
Aluminum bottle
Glass bottle
Poly bottle
Poly pail
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.25g/mL
pH Neutral
Boiling Point 290°C (1013 hPa)
Flash Point 160°C (320°F)
Ignition Point 698°F
Odor Odorless
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 500 mL
UN Number NONH for all modes of transport
Beilstein 635685
Solubility Information Completely miscible in water
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'