Learn More
Glycerol, 99.0 to 101.0%, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
47.89 € - 56.57 €
Chemical Identifiers
CAS | 56-81-5 |
---|---|
Molecular Formula | C3H8O3 |
Molecular Weight (g/mol) | 92.094 |
MDL Number | MFCD00004722 |
InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
PubChem CID | 753 |
ChEBI | CHEBI:17754 |
IUPAC Name | propane-1,2,3-triol |
SMILES | C(C(CO)O)O |
Description
Used for long-term storage of bacterial cultures. Step gradients for bacteriophage/particle purification. Mammalian cell transfection. Storage buffers for enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used for long-term storage of bacterial cultures
- Step gradients for bacteriophage/particle purification
- Mammalian cell transfection
- Storage buffers for enzymes
Chemical Identifiers
56-81-5 | |
92.094 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
753 | |
propane-1,2,3-triol |
C3H8O3 | |
MFCD00004722 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
CHEBI:17754 | |
C(C(CO)O)O |
Specifications
56-81-5 | |
1.26 | |
160°C | |
HOCH2CH(OH)CH2OH | |
MFCD00004722 | |
1.26 | |
PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
propane-1,2,3-triol | |
753 | |
92.09 | |
Molecular Biology, Ultrapure | |
Glycerol |
18°C | |
182°C (20 mmHg) | |
C3H8O3 | |
1.470-1.475 | |
635685 | |
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
C(C(CO)O)O | |
92.094 | |
CHEBI:17754 | |
99.0 to 101.0% | |
Viscous Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 200-289-5
missing translation for 'rtecsNumber' : MA8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.