missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, 100% Vegetable Based, U.S.P., J.T. Baker™

194.00 € - 1915.00 €

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15587974
View Documents Promotion Details
Avantor J.T.Baker
2142-01
500 mL Narrow Mouth Amber Glass Bottle
194.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15597974
View Documents Promotion Details
Avantor J.T.Baker
2142-03
4 L Narrow Mouth Amber Glass Bottle
928.00 €
4 L
Please to purchase this item. Need a web account? Register with us today!
15507984
View Documents Promotion Details
Avantor J.T.Baker
2142-07
19 L Poly Pail
1915.00 €
19 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO

Specifications

56-81-5
C3H8O3
1L = 1.26kg
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.1
USP
199.00°C
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
CHEBI:17754
100% Vegetable Based
Glycerin
Videos
SDS
Documents

Documents

Product Certifications