Learn More
Glutaryl dichloride, 97%, Thermo Scientific Chemicals
54.27 € - 150.75 €
Chemical Identifiers
CAS | 2873-74-7 |
---|---|
Molecular Formula | C5H6Cl2O2 |
Molecular Weight (g/mol) | 169.01 |
MDL Number | MFCD00000755 |
InChI Key | YVOFTMXWTWHRBH-UHFFFAOYSA-N |
Synonym | glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride |
PubChem CID | 17887 |
IUPAC Name | pentanedioyl dichloride |
SMILES | C(CC(=O)Cl)CC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10102870
|
Thermo Scientific Acros
119990250 |
25 mL | Glass bottle |
54.27 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676112
|
Thermo Scientific Acros
119991000 |
100 mL | Glass bottle |
150.75 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2873-74-7 | |
169.01 | |
YVOFTMXWTWHRBH-UHFFFAOYSA-N | |
17887 | |
C(CC(=O)Cl)CC(=O)Cl |
C5H6Cl2O2 | |
MFCD00000755 | |
glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride | |
pentanedioyl dichloride |
Specifications
2873-74-7 | |
1.3240g/mL | |
106°C | |
97% | |
ClCO(CH2)3COCl | |
MFCD00000755 | |
1.324 | |
Solubility in water: reacts with water. | |
C(CC(=O)Cl)CC(=O)Cl | |
169.01 | |
169.01 | |
Liquid |
Brown-Red or Colorless to Yellow | |
216.0°C to 218.0°C | |
Authentic | |
C5H6Cl2O2 | |
1.4720 to 1.4760 | |
02, 634 | |
glutaryl dichloride, glutaryl chloride, glutaroyl chloride, glutaric acid dichloride, 1,3-bis chlorocarbonyl propane, glutaroyl dichloride, glutaryl chloride 7ci,8ci, glutaryldichloride, glutarylchloride, glutaric acid chloride | |
YVOFTMXWTWHRBH-UHFFFAOYSA-N | |
pentanedioyl dichloride | |
17887 | |
97% | |
Glutaryl dichloride, 97% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-711-1
missing translation for 'rtecsNumber' : SA2583000
missing translation for 'tsca' : TSCA
- Glutaryl dichloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.