Learn More
Glutaraldehyde, 25% aq. soln., Thermo Scientific Chemicals
26.13 € - 91.46 €
Chemical Identifiers
CAS | 111-30-8 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00007025 |
InChI Key | SXRSQZLOMIGNAQ-UHFFFAOYSA-N |
Synonym | glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde |
PubChem CID | 3485 |
ChEBI | CHEBI:64276 |
IUPAC Name | pentanedial |
SMILES | O=CCCCC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448900
|
Thermo Scientific Alfa Aesar
A17876.AE |
100 mL |
26.13 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423488
|
Thermo Scientific Alfa Aesar
A17876.AP |
500 mL |
28.24 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433488
|
Thermo Scientific Alfa Aesar
A17876.0F |
2500 mL |
91.46 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker and as a disinfectant for medical equipment. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens. It is a disinfectant, which is rapidly effective against vegetative forms of Gram-positiveand Gram-negative bacteria.
Solubility
Miscible with water, ethanol, benzene, ether, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.
Notes
Heat and air sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents.
Chemical Identifiers
111-30-8 | |
100.12 | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
3485 | |
pentanedial |
C5H8O2 | |
MFCD00007025 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
CHEBI:64276 | |
O=CCCCC=O |
Specifications
111-30-8 | |
0.25 | |
OHC(CH2)3CHO | |
MFCD00007025 | |
605390 | |
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
pentanedial | |
3485 | |
100.12 | |
Glutaraldehyde |
1.062 | |
C5H8O2 | |
1.373 | |
UN2922 | |
14,4472 | |
Miscible with water,ethanol,benzene,ether,acetone,dichloromethane,ethylacetate,isopropanol,n-hexane and toluene. | |
O=CCCCC=O | |
100.12 | |
CHEBI:64276 | |
25% aq. soln. |
Safety and Handling
GHS H Statement
H331-H334-H314-H302-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302+H332-H314-H317-H334
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 203-856-5
missing translation for 'rtecsNumber' : MA2450000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Glutaraldehyde, 25% aq. soln.
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only