Learn More
DWK Life Sciences DURAN™ Short Ground Socket, finished
For connections where space is at a premium.
Brand:  DWK Life Sciences 286531801
Product Code. 15864808
  • 213.00 € / Pack of 50
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The short-section version of the DURAN™ sleeves is ideal for use where space is limited and tightness requirements are lower.
  • According to DIN 12249 Form E
  • Ground (finer grinding quality, less roughness depth)
  • The finer surface reduces the risk of jamming the cuts.
  • Minimized grease requirement
  • Very low leakage rate
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Installation in glass apparatus and devices
17 mm
1.8 mm