missing translation for 'onlineSavingsMsg'
Learn More

Rogo Sampaic™ Glass Lames

Specifications

Material Glass
Length (Metric) 76 mm
Width (Metric) 26 mm
Color White
Quantity 50 Pack
View More Specs

Description

Description

Specifications

Specifications

Glass
26 mm
50 Pack
76 mm
White
76 x 26 mm
Videos
SDS
Documents

Documents

Product Certifications