missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Glass Fiber Pre-Filters
Sartorius Stedim Biotech™ Glass Fiber Pre-Filters are suitable for liquid filtration, prefiltration, and analytical sample preparation.
Brand: Sartorius Stedim Biotech™ 13400--45------Q
This item is not returnable.
View return policy
Description
- Primarily used as a pre-filter
- Hydrophilic
- Glass microfiber with binder
- Non-sterile, bulk packed
- White, w/o grid
- Manufactured with synthetic binding agents to ensure that the filter has a defined strength
- Mechanically and chemically stable
Specifications
73 g/m2 | |
Glass Microfiber with Binder | |
Hydrophilic | |
180°C |
Disc | |
13400 | |
500/Pk. | |
45 mm |