missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Glass Fiber Pre-Filters

Sartorius Stedim Biotech™ Glass Fiber Pre-Filters are suitable for liquid filtration, prefiltration, and analytical sample preparation.

Brand:  Sartorius Stedim Biotech™ 13400--80------N

 View more versions of this product

Product Code. 15902480

  • 95.10 € / Pack of 100
Estimated Shipment: 06-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Primarily used as a pre-filter
  • Hydrophilic
  • Glass microfiber with binder
  • Non-sterile, bulk packed
  • White, w/o grid
  • Manufactured with synthetic binding agents to ensure that the filter has a defined strength
  • Mechanically and chemically stable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

73 g/m2
Glass Microfiber with Binder
Hydrophilic
180°C
Disc
13400
100/Pk.
80 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers