Learn More
Poulten Graf™ Glass BOD Bottle
Prevents air entrapment for your biological oxygen demand needs. Poulten Graf™ Glass BOD Bottle is made of soda lime Volac™ glass with an interchangeable stopper.
Brand:  Poulten Graf™ GS748/A
Product Code. 10618212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Glass peg stopper adjusted to leave a "well" for water seal at bottle opening
  • Pointed cone on stopper to prevent air entrapment
  • Manufactured from clear glass
Specifications
Specifications
Glass