missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ ScienceLine Glass Single Reference Electrode

Especially useful for the separation of combination electrodes into separate measuring and reference electrodes. SI Analytics™ ScienceLine Glass Single Reference Electrode are for special applications and make complex applications even more reliable.

330.00 €

Specifications

Description Ceramic Diaphragm
Material Glass
pH Range 0 to 14
Type Reference Electrode
View More Specs

Products 1
Product Code Brand Reference Electrolyte Diameter (Metric) Body Length (Metric) Price Quantity & Availability  
Product Code Brand Reference Electrolyte Diameter (Metric) Body Length (Metric) Price Quantity & Availability  
10213462
View Documents Promotion Details
SI Analytics™
1070048
Calomel 10 mm 103 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • The combination of pure measuring electrodes for i.e. ion-selective measurements or special constructions with one reference electrode
  • Redox titrations with a constant pH value with glass electrodes as reference and redox single sensors as measuring electrode
  • Application of double-electrolyte electrodes for an optimal adaptation of reference electrolyte to the measuring media.
Specifications

Specifications

Ceramic Diaphragm
Glass
0 to 14
Reference Electrode
Videos
SDS
Documents

Documents

Product Certifications