Learn More
Gilson™ Safe Aspiration Station
Vacuum-Controlled Safe Aspiration Station with small footprint.
Description
- Closed, stand-alone unit designed to prevent contamination hazards
- Compact design with adjustable speed allowing quick aspiration
- Always ready to use by simply pressing the handle button
- Handle accommodates different types of adaptors to aspirate from a variety of vessels
- Convenient coupling for quick bottle emptying
- In-line hydrophobic filter traps aerosols
- Connect two handles to the station with the Y-shape connector
- Motor starts only when the vacuum pressure drops
- Level detection prevents overflow
Specifications
Clear | |
1 Ea. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Gilson™ Safe Aspiration Station