Learn More
Gibco™ MEM α, no nucleosides
MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells.
78.50 €
Specifications
Classification | Animal Origin-free |
---|---|
Concentration | 1 X |
Form | Liquid |
Product Type | MEM α (Minimum Essential Medium α) |
Serum Level | Standard Serum Supplementation |
Requires: Supplementation
Description
MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This MEM α is modified as follows:
With: Phenol Red, L-glutamine
Without: Ribonucleosides, Deoxyribonucleosides
Using DMEM α
MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR-negative cells. This product is made with Earle's salts. MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature
Specifications
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
cGMP-compliant under the ISO 13485 standard |
1 X | |
MEM α (Minimum Essential Medium α) | |
Sterile-filtered | |
Gibco™ |
Safety and Handling
missing translation for 'shelfLife' : 12 Months From Date of Manufacture
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.