Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, USDA-approved regions
Important Update: We've recently introduced Value Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified USDA origin and Brazil (South American) origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ A3840201
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.
- Replacing with Fetal Bovine Serum, value
- Sera application: for cell culture with robust cell lines, good value for basic research
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Origin: countries meeting USDA importation requirements
Gibco serum delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Fetal | |
Countries meeting USDA importation requirements | |
One Shot | |
50 mL | |
Sterile | |
Triple-filtered, 0.1μm | |
One Shot™ |
Fetal Bovine Serum, heat inactivated, qualified, One Shot™, USDA-approved regions | |
Bovine | |
Liquid | |
Fetal Bovine Serum | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
Qualified | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.