missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giantin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22270-1-AP-20UL
This item is not returnable.
View return policy
Description
Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.Specifications
Giantin | |
Polyclonal | |
Unconjugated | |
GOLGB1 | |
GCP, GCP372, giantin, GOLGB1, Golgin subfamily B member 1, GOLIM1, MacroGolgin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2804 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14789 | |
GOLGB1 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction