missing translation for 'onlineSavingsMsg'
Learn More

Giantin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 22270-1-AP-150UL

 View more versions of this product

Product Code. 16821815

  • 410.00 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Golgin subfamily B member 1 (GOLGB1) is a single pass type I membrane protein located in the golgi apparatus membrane. GOLGB1 may participate in forming intercisternal cross-bridges of the Golgi complex.
Specifications

Specifications

Giantin
Polyclonal
Unconjugated
GOLGB1
GCP, GCP372, giantin, GOLGB1, Golgin subfamily B member 1, GOLIM1, MacroGolgin
Rabbit
Antigen Affinity Chromatography
RUO
2804
-20°C
Liquid
Immunocytochemistry, Immunofluorescence
0.54 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q14789
GOLGB1
Peptide
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.